ESD safety ankle boots, lace-up, A515
Supplier: ATLAS - the shoe company
24400-36 24400-37 24400-38 24400-39 24400-40 24400-41 24400-42 24400-43 24400-44 24400-45 24400-46 24400-47 24400-48 24400-49
114-8461EA 0 EUR
114-8461 114-8462 114-8463 114-8464 114-8465 114-8466 114-8467 114-8468 114-8469 114-8470 114-8471 114-8472 114-8473 114-8474
ESD safety ankle boots, lace-up, A515
Shoes and Boots
Sneaker style ESD safety shoes made from full-grained calf leather and Sneaker line sole.
  • Skin friendly, abrasion resistant active-X functional lining for enhanced breathability
  • Interchangeable insole
  • 3D cushioning system: Pressure-relieving insole, cushioning comfort midsole made from MPU, dynamic outsole system
  • Metal-free penetration resistant sole
  • Protective toe cap (200 J)
  • Electrical contact resistance of 35 mega Ohm
Colour: Brown
Certifications: EN 6130 (ESD); DGUV 112-191; EN ISO 20345; S3