Occupational shoes, Velcro® fastening, Linz
Supplier: LIEFERSERVICE UND LOGISTIK
583174/45 583174/44 583174/43 583174/42 583174/41 583174/40 583174/39 583174/38 583174/37 583174/36 583174/46
114-8507EA 135 EUR
114-8507 114-8506 114-8505 114-8504 114-8503 114-8502 114-8501 114-8500 114-8499 114-8498 114-8508
Occupational shoes, Velcro® fastening, Linz
Shoes and Boots
Clog made from comfortable unsplit smooth leather upper, an adjustable backstrap with a hook-and-loop fastene and a nonslip, oil- and grease-resistant super grip sole.
  • Provides reliable protection from impact injuries
  • Easy to clean
  • Anatomically shaped cork-latex footbed
  • Midsole made from EVA provides additional cushioning
  • Sole promotes the ideal rolling motion of the foot
Colour: White
Certifications: EN ISO 20347:2012, SRC