UHPLC columns, InfinityLab Poroshell 120
Supplier: Agilent
693975-901 699775-901 693775-901 699975-901 699975-301 695975-901 695975-301 695775-901 693975-301 699770-901 699770-901K 693770-901K 693970-301K 693970-901K 699970-901 699970-901K 695770-901 695770-901K 695970-301 695970-301K 695970-901 695970-901K 699970-301 699970-301K 650750-901K 690970-301K 820750-926 821725-926 823750-926 690970-901K 823750-940 820750-920 821725-919 820750-918 820750-919 821725-918 821725-920 823750-917 823750-918 823750-919 823750-920 820750-917 821725-917 693675-912 693675-408 693675-901 693675-902 693675-906 695675-408 695675-901 695675-906 695675-912 821725-940 821725-941 821725-942 821725-943 821725-944
HEWL693975-901EA 999 EUR
HEWL693975-901 HEWL699775-901 HEWL693775-901 HEWL699975-901 HEWL699975-301 HEWL695975-901 HEWL695975-301 HEWL695775-901 HEWL693975-301 HEWL699770-901 HEWL699770-901K HEWL693770-901K HEWL693970-301K HEWL693970-901K HEWL699970-901 HEWL699970-901K HEWL695770-901 HEWL695770-901K HEWL695970-301 HEWL695970-301K HEWL695970-901 HEWL695970-901K HEWL699970-301 HEWL699970-301K HEWL650750-901K HEWL690970-301K HEWL820750-926 HEWL821725-926 HEWL823750-926 HEWL690970-901K HEWL823750-940 HEWL820750-920 HEWL821725-919 HEWL820750-918 HEWL820750-919 HEWL821725-918 HEWL821725-920 HEWL823750-917 HEWL823750-918 HEWL823750-919 HEWL823750-920 HEWL820750-917 HEWL821725-917 HEWL693675-912 HEWL693675-408 HEWL693675-901 HEWL693675-902 HEWL693675-906 HEWL695675-408 HEWL695675-901 HEWL695675-906 HEWL695675-912 HEWL821725-940 HEWL821725-941 HEWL821725-942 HEWL821725-943 HEWL821725-944
UHPLC columns, InfinityLab Poroshell 120
These InfinityLab Poroshell 120 1,9 µm columns provide high performance under the highest pressures.
  • Ultra-fast separations increase productivity
  • High resolution separations increase the accuracy and precision of analysis results, reducing the need for re-work
Stability for high-pH mobile phases
InfinityLab Poroshell HPH-C18 particles are made by chemically modifying the porous layer using proprietary technology that provides high-pH stability. That means you can use InfinityLab Poroshell 120 1,9 µm columns for all your fast LC method development needs - regardless of mobile phase pH.
Rugged performance: Column lifetime test at 1000 bar
InfinityLab Poroshell 120 1,9 µm columns remain stable for 5000 injections under high pressure UHPLC conditions. That’s at least five times longer than other superficially porous (SPP) columns.