4-Hydroxymethyl-2,6,7-trioxa-1-phosphabicyclo[2.2.2]octane-1-oxide 98%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: 2,6,7-Trioxa-1-phosphabicyclo[2.2.2]octane-4-methanol 1-oxide
H61917.06 H61917.14
H61917.06EA 91.3 EUR
H61917.06 H61917.14
4-Hydroxymethyl-2,6,7-trioxa-1-phosphabicyclo[2.2.2]octane-1-oxide 98%
4-Hydroxymethyl-2,6,7-trioxa-1-phosphabicyclo[2.2.2]octane-1-oxide
Formula: C₅H₉O₅P
MW: 180.096 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01677612
CAS Number: 5301-78-0
Order Now
Specification Test Results
Assay 97.5%