Pt-Co/Hazen/APHA colour reference standards, ASTM D 1209, Supelco®
Supplier: Merck
134140-500ML
134150-500ML
134160-500ML
134170-500ML
134180-500ML
134190-500ML
SIAL134140-500MLEA
242
EUR
SIAL134140-500ML
SIAL134150-500ML
SIAL134160-500ML
SIAL134170-500ML
SIAL134180-500ML
SIAL134190-500ML
Pt-Co/Hazen/APHA colour reference standards, ASTM D 1209, Supelco®
Standards
Colour Reference and Spectroscopy Standards
Certifications: These products are certified reference materials (CRM) certified under ISO 9001 Quality system.