Reagent reservoirs
Supplier: HEATHROW
120644 120645 120648 120649 120690 120652 120651 120646 120647 120639 120640 120638 120642 120650 120637 120641 120643
HEAT120644EA 176 EUR
HEAT120644 HEAT120645 HEAT120648 HEAT120649 HEAT120690 HEAT120652 HEAT120651 HEAT120646 HEAT120647 HEAT120639 HEAT120640 HEAT120638 HEAT120642 HEAT120650 613-6915 HEAT120641 HEAT120643
Reagent reservoirs
Reagent Reservoirs
These reagent reservoirs offer a fresh and modern design with enhanced features to streamline pipetting procedures. They are designed for multi-channel pipettes and accommodate most common brands. A full range of volumes and styles is available to accommodate required volume of reagents and reduce waste.
  • 'V' shaped well design allows maximum reagent recovery
  • Easy to read graduation markings located on the top edge with a fill line on inner walls to indicate volume levels
  • Drip-free pour-off spouts at every corner for spill-free pouring
  • A wide base ensures stability and tiered wall design prevents deformation when handling reducing the chance of spills
  • A unique nesting design allows easy separation that reduces the chance of warped or damaged product
  • Dual-reservoir design allows pipetting of two reagents and/or maximises recovery of small samples
The reservoirs are free of animal products and manufactured in a cleanroom with sterile versions certified as RNase-/DNase- and pyrogen-free.
Certifications: Quality certificate available.
Ordering information: Available in polystyrene (PS) or polyvinyl chloride (PVC). Bulk and five pack versions come conveniently stacked to help save space when storing.
Packaging: Lot numbers indicated on sterile versions. Easy tear packaging makes handling quick and efficient.