Titrimetric and colorimetric rapid tests, oxygen, VISOCOLOR®
Supplier: MACHEREY-NAGEL
915209 931088 931288 915009
MANA915209EA 82.7 EUR
MANA915209 MANA931088 MANA931288 MANA915009
Titrimetric and colorimetric rapid tests, oxygen, VISOCOLOR®
Environmental Test Kits
The VISOCOLOR® testkits are devided into VISOCOLOR® alpha, ECO and HE. Every product range offering its specific strenght to provide a tailored test for each and every user and application.
  • Portable
  • Environmentally friendly
  • Convenient handling
  • Application-oriented measuring ranges
The solubility of oxygen in water depends on the temperature, the pressure and the other components of the water. The oxygen content of a water at the time of sampling is often quoted as the percentage of the possible saturation.
Ordering information: Please note: When ordering test kit 931088 for the first time, you also need an oxygen bottle, REF 915 498.