Examination gloves, Nitrile, Nitril® NextGen®
Supplier: MEDITRADE
1283A-S_U 1283A-L_U 1283A-XL_U 1283A-M 1283ALPALETT_U 1283AMPALETT_U
ROES1283A-S_UEA 53.3 EUR
ROES1283A-S_U ROES1283A-L_U ROES1283A-XL_U ROES1283A-M ROES1283ALPALETT_U ROES1283AMPALETT_U
Examination gloves, Nitrile, Nitril® NextGen®
Gloves
The disposable Nitrile® NextGen® gloves are made from nitrile butadiene rubber (NBR). The 'Innovation-Soft-Formula' gives this nitrile glove an almost latex-like, high elasticity and tear resistance. The polymer inner coating allows effortless donning.
  • Intuitive fit with rolled edge
  • Non sterile
  • Ambidextrous
  • Micro-roughened
  • For single use
Nitrile is free of allergenic proteins, therefore suitable for users with diagnosed latex allergy. Nitrile is free from most excipients that can cause Type IV allergies.
Suitable for cleaning agents and disinfection and for other applications in the laboratory, in the pharmaceutical industry, for professional cleaning, in catering and for handling food in general. Ideal for sensitive users with diagnosed latex allergy (Type I).
Certifications: EN 455, EN 374-1/2/3; Cat. III; AQL 1,5
Food contact approved