Ammonium acetate 5 M in aqueous solution for molecular biology
Supplier: Thermo Fisher Scientific
Warning
Synonyms: Acetic acid ammonium salt
J60688.AD J60688.AE
J60688.ADEA 53.4 EUR
J60688.AD J60688.AE
Ammonium acetate 5 M in aqueous solution for molecular biology
Ammonium acetate
Ammonium acetate is used for routine precipitation of nucleic acids, and is especially useful for reducing the coprecipitation of unwanted dNTPs and contaminating oligosaccharides.
It is used as an important reagent for studying molecular biology, biological buffers, reagents and DNA and RNA purification. It is a popular buffer for mobile phases for HPLC with ESLD detection, for ESI mass spectrometry of proteins and other molecules, and has been used to replace cell buffers with non-volatile salts. Ammonium acetate is also used in protein studies and protein preparation. It can be used in the protein purification steps of dialysis to remove contaminants through diffusion and, when combined with distilled water, as a protein precipitating agent. In organic chemistry, ammonium acetate solution is useful as a catalyst in the knoevenagel condensation and as a source of ammonia in the borch reaction. Additionally, it is occasionally used commercially as a biodegradable de-icing agent and as an additive in food as an acidity regulator.
Formula: H₃CCOONH₄
MW: 77.08 g/mol
Melting Pt: 114
Flash Pt: 136
Storage Temperature: Freezer
MDL Number: MFCD00013066
CAS Number: 631-61-8
EINECS: 211-162-9
Merck Index: 13,00495
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd160345483553318b94053de396335a9b18425f&vid=c6375b539008d6b43f50408d30b61261444faebc&prid=web&clid=SAPDAM