Beverage refrigerators, FKU series
Supplier: Liebherr
FKU1800 FKU1805 FKU1802
472-2181EA 739 EUR
472-2181 472-2182 LIEBFKU1802
Beverage refrigerators, FKU series
Refrigerators
These multi-purpose refrigerators with static cooling and three shelves can be built under counters or workbenches. Ideal for cooling beverages or other non hazardous liquids in bottles.
  • Adjustable stainless steel feet
  • Automatic defrost; mechanical temperature control
  • FKU 1805: Digital temperature display indicates the refrigerator’s interior temperature on the outside
  • FKU 1803: LED lighting, with separate switch
  • Environmentally friendly refrigerants (R600a and R290)
  • Reversible door hinges (left/right)
  • Available with stainless steel or insulating glass door
Temperature range: +2...+10 °C
Capacity: 160 l (115 × 0,5 l PET bottles or 217 × 0,33 l cans)