Ferroin (1,10-phenanthrolineferrous sulphate complex) 1/40 mol/l, AVS TITRINORM redox indicator
Supplier: VWR Chemicals
Synonyms: Tris(1,10-phenanthroline)iron (II) sulphate, Tris(1,10-phenanthroline-N1,N10)iron sulphate
86180.180EA 56.7 EUR
86180.180 86180.260
Ferroin (1,10-phenanthrolineferrous sulphate complex) 1/40 mol/l, AVS TITRINORM redox indicator
Ferroin (1,10-Phenanthroline-ferrous sulphate-complex)
Formula: C₃₆H₂₄FeN₆SO₄
MW: 692.54 g/mol
Density: 0.9999
MDL Number: MFCD00036428
CAS Number: 14634-91-4
Order Now
Specification Test Results
Spec. Absorptivity A 1%/1cm (λmax; 0.003 %; buffer pH 7.0) 180 - 200