Sterile sleeve/glove system, BioClean™
Supplier: Nitritex
GSG10NIT80 GSG10NIT85
112-6122EA 6360 EUR
112-6122 112-6123
Sterile sleeve/glove system, BioClean™
Gloves
Clean and sterile sleeve/glove system, nitrile sleeve attached to a hand specific polychloroprene (BioClean BPZS) glove by a channel ring and O-ring.
  • Sleeve and glove tested against ASTM D6978-05 for handling chemo drugs
  • Ultra-clean surface ensures product protection
  • Sleeve 100% inspection and air leak tested (prior to being guillotined)
  • Sleeve suitable for autoclaving
  • Gamma irradiated (Sterility Assurance Level (SAL) of 10⁻⁶)
  • Sleeve can be sanitised by VHP or IPA
Compatible with ISO Class 4 & EU GMP Grade A cleanrooms.
* Length of the completeMore Product Information
Order Now
SPECIFICATIONS
Ambidextrous / hand-specific Hand-specific
Environmentally Preferable
Disposable / reusable Reusable
Latex / latex-free Latex-free
Powdered / powder-free Powder-free
Cleanroom class 10/ISO 4; EU GMP (A)
Material Nitrile
Finish Smooth
Cuff style Beaded
Cuff thickness 0,11 mm
Finger thickness 0,18 mm
Palm thickness 0,15 mm
EN norm EN ISO 374-1 Type B (KOT), EN ISO 374-5 VIRUS, EN 421
Sterility Sterile