VWR® LAG Advanced, Analytical Balances with Graphic Display
Supplier: VWR Collection
611-3267EA 1600 EUR
611-3267 611-3268 611-3297 611-3376 611-3457 611-4302
VWR® LAG Advanced, Analytical Balances with Graphic Display
Balances and Scales
An economical range of reliable, high performance analytical balances. All models have a stainless steel top pan.
  • Check weighing (alarm function), weight totalising, density determinations of samples and piece counting functions
  • Models have large graphic display with added functionality for formulation, GLP, date and time, multi-language
  • Option of external or internal calibration, RS232 included as standard
  • Weighing chamber L×W×H: 162×171×225 mm
User can choose three options for models with internal calibration (denoted by 'i' at end of the model name):
Balance will calibrate using the internal weight by pressing CAL button
Balance will automatically recalibrate when ambient temperature changes and/or after a certain time has passed (Incal+)
The user can choose to calibrate the balance with an external weight.
Certifications: EN 61326-1:2013, EN 61010-1: 2010, EN 61010-2-010:2014, EN 50581:2012.
Accessories information: Not suitable for use with tablet systems.
Ordering information: For more information on the full range of accessories, including printers, software, density control kit for the LAG series, external keyboard and PC connection cable, please visit our website or contact your local Avantor sales office.
The balances are also available with a factory calibration certificate. Please contact Avantor for more information about this option.
Delivery information: Supplied with mains adapter, universal plug and plastic in-use protective cover. Calibration weights are available from Avantor. Please see 'Calibration weights'.
Caution: Models with a large graphic display are denoted by G in the model name. Similarly i shows internal calibration. Models with suffix -C are supplied with a factory calibration certificate. See website for details of the full range of models.