ESD safety shoes, lace-up, uvex 1 sport, 6593
Supplier: UVEX
6593.7/49 6593.7/35 6593.7/36 6593.7/37 6593.7/38 6593.7/39 6593.7/40 6593.7/41 6593.7/42 6593.7/43 6593.7/44 6593.7/45 6593.7/46 6593.7/47 6593.7/48 6593.7/50 6593.7/51 6593.7/52 6593.8/35 6593.8/36 6593.8/37 6593.8/38 6593.8/39 6593.8/40 6593.8/41 6593.8/42 6593.8/43 6593.8/44 6593.8/45 6593.8/46 6593.8/47 6593.8/48 6593.8/49 6593.8/50 6593.8/51 6593.8/52
UVEX6593.7/49EA 107 EUR
UVEX6593.7/49 UVEX6593.7/35 UVEX6593.7/36 UVEX6593.7/37 UVEX6593.7/38 UVEX6593.7/39 UVEX6593.7/40 UVEX6593.7/41 UVEX6593.7/42 UVEX6593.7/43 UVEX6593.7/44 UVEX6593.7/45 UVEX6593.7/46 UVEX6593.7/47 UVEX6593.7/48 UVEX6593.7/50 UVEX6593.7/51 UVEX6593.7/52 UVEX6593.8/35 UVEX6593.8/36 UVEX6593.8/37 UVEX6593.8/38 UVEX6593.8/39 UVEX6593.8/40 UVEX6593.8/41 UVEX6593.8/42 UVEX6593.8/43 UVEX6593.8/44 UVEX6593.8/45 UVEX6593.8/46 UVEX6593.8/47 UVEX6593.8/48 UVEX6593.8/49 UVEX6593.8/50 UVEX6593.8/51 UVEX6593.8/52
ESD safety shoes, lace-up, uvex 1 sport, 6593
Shoes and Boots
The uvex 1 sport is both youthful and stylish combined with first-class wearer comfort, thanks to the lightweight design, climate-optimised high-tech materials, and ergonomically designed outsole.
  • Work shoes meet the ESD specifications with leakage resistance under 35 megaohms
  • Breathable textile upper
  • No toe cap
This modern, ultra-lightweight and flexible work shoe is made from synthetic materials, so it is suitable for people allergic to chrome. Sole materials are free from silicones, plasticisers, and other substances that interfere with wetting agents. The ergonomically designed outsole is made from dual-density polyurethane that offers excellent slip resistance.
Colour: Black
Certifications: Protection class O1 FO SRC in accordance with EN ISO 20347:2012; EN 61340 (ESD).