Sodium hypochlorite ∼ 1N (0.5 mol/l) in sodium hydroxide solution 0.1 mol/l low in bromine
Supplier: VWR Chemicals
Danger
230393LEA 92.4 EUR
230393L 230395N
Sodium hypochlorite ∼ 1N (0.5 mol/l) in sodium hydroxide solution 0.1 mol/l low in bromine
Sodium hypochlorite
Formula: ClNaO
MW: 74.44 g/mol
Melting Pt: -6
Density: 1 g/cm³ (20 °C)
MDL Number: MFCD00011120
CAS Number: 7681-52-9
EINECS: 231-668-3
UN: 1791
ADR: 8 II
Merck Index: 13,08702
Order Now
Specification Test Results
Sodium Hypochlorite (NaOCl) 0.9 - 1.1 N
Sodium Hydroxide 0.9 - 0.11 mol/l
Bromine (Br) >20 ppm