Florisil® 60-100 mesh for chromatography
Supplier: VWR Chemicals
Synonyms: Talc, Magnesium silicate, activated, Silicic acid magnesium salt, Florisil
24278.182EA 69.7 EUR
24278.182 24278.295 24278.230 24278.440
Florisil® 60-100 mesh for chromatography
Magnesium silicate
Formula: MgO₃Si
MW: 100.38 g/mol
MDL Number: MFCD00078249
CAS Number: 1343-88-0
EINECS: 215-681-1
Order Now
Specification Test Results
Identification Passes Test