Starch, soluble, AnalaR NORMAPUR® for iodometric titrations, analytical reagent
Supplier: VWR Chemicals
Synonyms: Starch, Vitex® iodine indicator, Amylodextrin
28610.291EA 387 EUR
28610.291 28610.187 28610.234
Starch, soluble, AnalaR NORMAPUR® for iodometric titrations, analytical reagent
Starch, soluble
Iodine indicator in iodometry
Formula: (C₆H₁₀O₅)n
MW: 162.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00082026
CAS Number: 9005-84-9
EINECS: 232-686-4
Merck Index: 13,08878
Order Now
Specification Test Results
Melting point 129 - 133 °C
Suited for iodine reagent Passes test