Zinc ≥98%, dust <10 μm, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
M-Clarity™ quality level = MQ200
209988-100G
209988-1KG
209988-5KG
SIAL209988-100GEA
56.6
EUR
SIAL209988-100G
SIAL209988-1KG
SIAL209988-5KG
Zinc ≥98%, dust <10 μm, Sigma-Aldrich®
Zinc
Formula:
Zn MW: 65.39 g/mol Boiling Pt: 908 (lit.) Melting Pt: 420 °C (lit.) Density: 7.13 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011291 CAS Number: 7440-66-6 EINECS: 231-175-3 UN: 3077 ADR: 9 III Merck Index: 12,10255 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...