Methanol-[D₄], Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: Methanol-[²H₄], Hydroxy methane-D4, Methanol-d4, Methyl-D3 alcohol-D, Carbinol-D4, Wood alcohol-D4, Methyl alcohol-D4, Tetradeuteromethanol

M-Clarity™ quality level = MQ200

417653-10G 417653-50G
SIAL417653-10GEA 183 EUR
SIAL417653-10G SIAL417653-50G
Methanol-[D₄], Sigma-Aldrich®
Methanol-[D4]
Formula: D₃COD
Boiling Pt: 65.3 °C (1013 hPa)
Melting Pt: –99 °C
Density: 0.888 g/cm³ (20 °C)
Flash Pt: 11 °C
Storage Temperature: Ambient
MDL Number: MFCD00044637
CAS Number: 811-98-3
EINECS: 212-378-6
UN: 1230
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR