Sodium hypochlorite (12% Cl₂) aqueous solution
Supplier: Roth Carl
Danger
9062.3 9062.2 9062.1
ROTH9062.3EA 26.3 EUR
ROTH9062.3 ROTH9062.2 ROTH9062.1
Sodium hypochlorite (12% Cl₂) aqueous solution
Sodium hypochlorite
Formula: ClNaO
MW: 74.44 g/mol
Melting Pt: –30…–20 °C
Density: 1.22…1.25 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00011120
CAS Number: 7681-52-9
EINECS: 231-668-3
UN: 1791
ADR: 8 II
Merck Index: 13,08702