DL-Cystine, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

C8630-1G C8630-5G
SIALC8630-1GEA 46.2 EUR
SIALC8630-1G SIALC8630-5G
DL-Cystine, Sigma-Aldrich®
DL-Cystine
Formula: C₆H₁₂N₂O₄S₂
Boiling Pt: 429 °C (1013 hPa)
Melting Pt: 227 °C
MDL Number: MFCD00084652
CAS Number: 923-32-0
EINECS: 213-094-5
Merck Index: 13,02811

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR