Fehling's reagent I (copper(II) sulphate solution) for qualitative determination of reducing sugars
Supplier: Thermo Fisher Scientific
Warning
Synonyms: Fehling A solution
270800010 270800025
ACRO270800010EA 44.6 EUR
ACRO270800010 ACRO270800025
Fehling's reagent I (copper(II) sulphate solution) for qualitative determination of reducing sugars
Fehling's reagent I (Copper (II) sulphate solution)
Formula: CuSO₄
MW: 159.61 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00010981
CAS Number: 7758-98-7
EINECS: 231-847-6
UN: 3082
ADR: 9 III
Order Now
Specification Test Results
Appearance Clear blue Solution
Solution contains 69.2 g CuSO4.5H2O/l H2O