Titanium(III) chloride 15% in hydrochloric acid 10%, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Titanium trichloride
M-Clarity™ quality level = MQ200
14010-1L-R 14010-250ML-R 14010-6X1L-R
SIAL14010-1L-REA 257 EUR
SIAL14010-1L-R SIAL14010-250ML-R SIAL14010-6X1L-R
Titanium(III) chloride 15% in hydrochloric acid 10%, Sigma-Aldrich®
Titanium (III) chloride
Formula: Cl₃Ti
MW: 154.24 g/mol
Boiling Pt: 960
Melting Pt: 440
Density: 1.32
MDL Number: MFCD00011266
CAS Number: 7705-07-9
EINECS: 231-728-9
UN: 1789
ADR: 8 II
Merck Index: 12,09620