Sodium chloride 5 mol/l (5 N) in aqueous solution, SAFC®
Supplier: Merck
Synonyms: Saline, NaCl, Sea salt, Halite
M-Clarity™ quality level = MQ500
59222C-1000ML 59222C-500ML
SIAL59222C-1000MLEA 54.7 EUR
SIAL59222C-1000ML SIAL59222C-500ML
Sodium chloride 5 mol/l (5 N) in aqueous solution, SAFC®
Sodium chloride
Formula: NaCl
MW: 58.44 g/mol
Boiling Pt: 1413
Melting Pt: 801
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
EINECS: 231-598-3
Merck Index: 14,08599