Magnesium sulphate 2.5 mol/l in aqueous solution, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
83266-100ML-F 83266-500ML-F
SIAL83266-100ML-FEA 86.7 EUR
SIAL83266-100ML-F SIAL83266-500ML-F
Magnesium sulphate 2.5 mol/l in aqueous solution, Sigma-Aldrich®
Magnesium sulphate
Formula: MgSO₄
Storage Temperature: Ambient
MDL Number: MFCD00011110
CAS Number: 7487-88-9
EINECS: 231-298-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1bc2ae189b0cbc1bfc599951528466503d0d9008&vid=63867221020ebd2c8c227492e609a4764639ff79&prid=web&clid=SAPDAM