Europium triacetate hydrate ≥99.999% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Fisher Scientific
Synonyms: Acetic acid europium (III) salt hydrate, Europium(III) acetate xhydrate, Europium (III) acetate hydrate
44056.09 44056.18
44056.09EA 274 EUR
44056.09 44056.18
Europium triacetate hydrate ≥99.999% (REO, rare earth oxide basis), REacton®
Europium triacetate hydrate
Purity: 99.999% CAS No.: 62667-64-5 Molecular Formula: Eu(OOCCH3)3.xH2O Formula Weight: 329.10(anhy) Physical Form: Lumps/Powder Sensitivity: Hygroscopic MDL No.: MFCD00150115
Formula: C₆H₉EuO₆·H₂O
MW: 347.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00150115
CAS Number: 62667-64-5
EINECS: 214-670-9