Ethyl-(S)-(-)-lactate, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Ethyl (S)-(-)-2-hydroxypropionate, Ethyl L(-)-lactate, (S)-Ethyl 2-hydroxypropanoate, (S)-(-)-Ethyl lactate
M-Clarity™ quality level = MQ100
69799-1L 69799-250ML
SIAL69799-1LEA 101 EUR
SIAL69799-1L SIAL69799-250ML
Ethyl-(S)-(-)-lactate, Sigma-Aldrich®
Ethyl (S)-(-)-lactate
Formula: C₅H₁₀O₃
MW: 118.13 g/mol
Boiling Pt: 154
Melting Pt: –25 °C
Density: 1.042 g/cm³ (20 °C)
Flash Pt: 53
Storage Temperature: Ambient
MDL Number: MFCD00004518
CAS Number: 687-47-8
EINECS: 211-694-1
UN: 1192
ADR: 3 III
Merck Index: 12,03850