Phosphotungstic acid hydrate, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Phosphotungstic acid hydrate, Wolframophosphoric acid hydrate, Dodecawolframophosphoric acid hydrate, 12-Tungstophosphate hydrate, Tungstophosphoric Acid Hydrate
M-Clarity™ quality level = MQ100
455970-10G 455970-50G
SIAL455970-10GEA 180 EUR
SIAL455970-10G SIAL455970-50G
Phosphotungstic acid hydrate, Sigma-Aldrich®
Phosphotungstic acid hydrate
Formula: H₃[P(W₃O₁₀)₄]·nH₂O
Melting Pt: 107 °C
MDL Number: MFCD00011342
CAS Number: 12501-23-4
EINECS: 215-682-7
UN: 3260
ADR: 8,III
Merck Index: 13,07450