Ethyl-(S)-(-)-lactate, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Ethyl (S)-(-)-2-hydroxypropionate, Ethyl L(-)-lactate, (S)-Ethyl 2-hydroxypropanoate, (S)-(-)-Ethyl lactate
M-Clarity™ quality level = MQ200
E34102-100G E34102-500G E34102-5G
SIALE34102-100GEA 37.7 EUR
SIALE34102-100G SIALE34102-500G SIALE34102-5G
Ethyl-(S)-(-)-lactate, Sigma-Aldrich®
Ethyl (S)-(-)-lactate
Formula: C₅H₁₀O₃
MW: 118.13 g/mol
Boiling Pt: 154
Melting Pt: –25 °C
Density: 1.042 g/cm³ (20 °C)
Flash Pt: 53
Storage Temperature: Ambient
MDL Number: MFCD00004518
CAS Number: 687-47-8
EINECS: 211-694-1
UN: 1192
ADR: 3 III
Merck Index: 12,03850