Methyl (S)-(-)-lactate ≥97%
Supplier: Thermo Fisher Scientific
|
Warning
|
Purity: 97%
CAS No.: 27871-49-4
Molecular Formula: C4H8O3
Formula Weight: 104.11
Flash Point: 49°(120°F)
Refractive Index: 1.4140
Melting Point: -44°
Boiling Point: 135-138°
Beilstein Reference: 1720587
Optical Rotation: -8.2° (neat)
UN No.: 3272
MDL No.: MFCD00064265
Formula:
C₄H₈O₃ Boiling Pt: 135…138 °C Melting Pt: –44 °C Density: 1.092 Flash Pt: 49 °C (120 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00064265 CAS Number: 27871-49-4 EINECS: 248-704-9 UN: 3272 ADR: 3,III |