Sodium sulphide hydrate ≥60%, scales, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: di-Sodium sulphide hydrate
M-Clarity™ quality level = MQ100
13468-1KG-R 13468-2.5KG-R 13468-6X1KG-R
SIAL13468-1KG-REA 65.6 EUR
SIAL13468-1KG-R SIAL13468-2.5KG-R SIAL13468-6X1KG-R
Sodium sulphide hydrate ≥60%, scales, Sigma-Aldrich®
Sodium sulphide hydrate
Formula: Na₂S·xH₂O
MW: 96.06 g/mol (anhy)
Boiling Pt: ∼920 °C (1013 hPa)
Melting Pt: 50
Density: ∼1.43 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00003498
CAS Number: 27610-45-3
EINECS: 215-211-5
UN: 1849
ADR: 8 II
REACH: 01-2119513694-38
Merck Index: 12,08829