Disodium metasilicate pentahydrate, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Sodium metasilicate pentahydrate
71746-1KG 71746-250G
SIAL71746-1KGEA 51.2 EUR
SIAL71746-1KG SIAL71746-250G
Disodium metasilicate pentahydrate, Sigma-Aldrich®
di-Sodium metasilicate pentahydrate
Formula: Na₂SiO₃·5H₂O
MW: 212.14 g/mol
Melting Pt: 72.2
Density: 1.749 g/cm³ (20 °C)
MDL Number: MFCD00149176
CAS Number: 10213-79-3
EINECS: 229-912-9
UN: 3253
ADR: 8 III
Merck Index: 13,08716