Copper(II) perchlorate hexahydrate, Sigma-Aldrich®
Supplier: Merck
Synonyms: Cupric perchlorate hexahydrate
M-Clarity™ quality level = MQ100
215392-100G 215392-5G
SIAL215392-100GEA 84.6 EUR
SIAL215392-100G SIAL215392-5G
Copper(II) perchlorate hexahydrate, Sigma-Aldrich®
Copper (II) perchlorate hexahydrate
Formula: Cl₂CuO₈·6H₂O
MW: 370.54
Melting Pt: 82
Density: 1.0491 g/cm³ (20 °C)
MDL Number: MFCD00149666
CAS Number: 10294-46-9
EINECS: 237-391-4
UN: 1481
ADR: 5.1 II