Zirconyl nitrate hydrate, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Zirconium(IV) oxynitrate hydrate, Zirconium(IV) dinitrate oxide hydrate, Zirconium dinitrate oxide hydrate
M-Clarity™ quality level = MQ200
346462-100G 346462-500G
SIAL346462-100GEA 89.4 EUR
SIAL346462-100G SIAL346462-500G
Zirconyl nitrate hydrate, Sigma-Aldrich®
Zirconyl nitrate hydrate
Formula: N₂O₇Zr·H₂O
MW: 249.25 g/mol
Density: 2.04 g/cm³ (20 °C)
MDL Number: MFCD00149896
CAS Number: 14985-18-3
EINECS: 237-529-3
UN: 1477
ADR: 5.1 III