(S)-(-)-1-Phenylethyl isocyanate, Sigma-Aldrich®

Supplier: Merck

Synonyms: (S)-(-)-α-Phenethyl isocyanate, (S)-(-)-α-Methylbenzyl isocyanate, (S)-1-phenylethylisocyanate, (S)-(-)-1-Phenethyl isocyanate

M-Clarity™ quality level = MQ100

220566-1G 220566-5G
SIAL220566-1GEA 60.9 EUR
SIAL220566-1G SIAL220566-5G
(S)-(-)-1-Phenylethyl isocyanate, Sigma-Aldrich®
(S)-(-)-1-Phenylethyl isocyanate
Formula: C₉H₉NO
Boiling Pt: 55 °C (1013 hPa)
Density: 1.05 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00002036
CAS Number: 14649-03-7
EINECS: 238-698-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR