Methyl acetoacetate, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
Methyl 3-oxobutanoate
M-Clarity™ quality level = MQ100
688886-1KG
688886-250KG
688886-25KG
SIAL688886-1KGEA
63.6
EUR
SIAL688886-1KG
SIAL688886-250KG
SIAL688886-25KG
Methyl acetoacetate, Sigma-Aldrich®
Methyl acetoacetate
Formula:
C₅H₈O₃ Boiling Pt: 170 °C (1013 hPa) Melting Pt: –39 °C Density: 1.076 g/cm³ (20 °C) Flash Pt: 67 °C |
MDL Number:
MFCD00008784 CAS Number: 105-45-3 EINECS: 203-299-8 Merck Index: 12,06090 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...