Palmitoyl chloride
Supplier: Merck
M-Clarity™ quality level = MQ200
P78-100ML
P78-1L
P78-500ML
SIALP78-100MLEA
95.6
EUR
SIALP78-100ML
SIALP78-1L
SIALP78-500ML
Palmitoyl chloride
Palmitoyl chloride
Formula:
C₁₆H₃₁ClO Boiling Pt: 324 °C (1013 hPa) Melting Pt: 12 °C Density: 0.91 g/cm³ (20 °C) Flash Pt: 182 °C |
MDL Number:
MFCD00000742 CAS Number: 112-67-4 EINECS: 203-996-7 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...