Geranyl butyrate, Sigma-Aldrich®

Supplier: Merck

Synonyms: Butyric acid geranyl ester, 3,7-Dimethylocta-2,6-dien-1-yl butyrate, Butanoic acid geranyl ester

M-Clarity™ quality level = MQ300

W251208-1KG-K W251208-9KG-K
SIALW251208-1KG-KEA 110 EUR
SIALW251208-1KG-K SIALW251208-9KG-K
Geranyl butyrate, Sigma-Aldrich®
Geranyl butyrate
Formula: C₁₄H₂₄O₂
MDL Number: MFCD00036538
CAS Number: 106-29-6
EINECS: 203-381-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR