Ethyl decanoate, Sigma-Aldrich®

Supplier: Merck

Synonyms: Ethyl caprate, Capric acid ethyl ester, Decanoic acid ethyl ester, Decylic acid ethyl ester

M-Clarity™ quality level = MQ100

148970-100ML 148970-500ML
SIAL148970-100MLEA 50 EUR
SIAL148970-100ML SIAL148970-500ML
Ethyl decanoate, Sigma-Aldrich®
Ethyl decanoate
Formula: C₁₂H₂₄O₂
Boiling Pt: 245 °C (1013 hPa)
Melting Pt: –20 °C
Density: 0.862 g/cm³ (20 °C)
MDL Number: MFCD00009581
CAS Number: 110-38-3
EINECS: 203-761-9
Merck Index: 13,03811

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR