Methyl decanoate, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ300
W505501-100G
W505501-1KG
W505501-SAMPLE
SIALW505501-100GEA
82
EUR
SIALW505501-100G
SIALW505501-1KG
SIALW505501-SAMPLE
Methyl decanoate, Sigma-Aldrich®
Methyl decanoate
Formula:
C₁₁H₂₂O₂ Boiling Pt: 224 °C (1013 hPa) Melting Pt: –18 °C Density: 0.8724 g/cm³ (20 °C) Flash Pt: 94 °C |
MDL Number:
MFCD00009580 CAS Number: 110-42-9 EINECS: 203-766-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...