Ethyl enantate, Sigma-Aldrich®
Supplier: Merck
Synonyms: Heptylic acid ethyl ester, Enanthic acid ethyl ester, Heptanoic acid ethyl ester, Ethyl heptanoate
M-Clarity™ quality level = MQ300
W243701-1KG-K W243701-20KG-K W243701-4KG-K
SIALW243701-1KG-KEA 85.5 EUR
SIALW243701-1KG-K SIALW243701-20KG-K SIALW243701-4KG-K
Ethyl enantate, Sigma-Aldrich®
Ethyl enantate
Formula: C₉H₁₈O₂
Boiling Pt: 372 °C (1013 hPa)
Melting Pt: –66 °C
Density: 0.868 g/cm³ (20 °C)
MDL Number: MFCD00009538
CAS Number: 106-30-9
EINECS: 203-382-9
Merck Index: 13,03867