Dimethyl succinate, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ100
112755-100G 112755-500G
SIAL112755-100GEA 46 EUR
SIAL112755-100G SIAL112755-500G
Dimethyl succinate, Sigma-Aldrich®
Dimethyl succinate
Formula: C₆H₁₀O₄
Boiling Pt: 200 °C (1013 hPa)
Melting Pt: 18…19 °C
Density: 1.117 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008466
CAS Number: 106-65-0
EINECS: 203-419-9
Merck Index: 12,09038