Diethylenetriaminepentaacetic acid, Supelco®
Supplier: Merck
Warning
Synonyms: N-Carboxymethyliminobis(ethylenenitrilo)tetra(acetic acid), Kestranal® Aza Pa, DTPA, Titriplex® V, Idranal V, Diethylenetriaminepentaacetic acid, 2,2',2'',2'''-((((Carboxymethyl)azanediyl)bis(ethane-2,1-diyl))bis(azanetriyl))tetraacetic acid, Diethylenetriamine-N,N,N',N'',N''-pentaacetic acid
M-Clarity™ quality level = MQ100
32319-100G-F 32319-500G-F
SIAL32319-100G-FEA 73.4 EUR
SIAL32319-100G-F SIAL32319-500G-F
Diethylenetriaminepentaacetic acid, Supelco®
Diethylenetriaminepentaacetic acid
Formula: C₁₄H₂₃N₃O₁₀
Melting Pt: 220…222 °C
Density: 1.558 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004289
CAS Number: 67-43-6
EINECS: 200-652-8
UN: 3077
ADR: 9,III
Merck Index: 13,07202