Urethane, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Carbamic acid ethyl ester, Ethyl carbamate
M-Clarity™ quality level = MQ200
U2500-100G U2500-250G U2500-500G
SIALU2500-100GEA 62.7 EUR
SIALU2500-100G SIALU2500-250G SIALU2500-500G
Urethane, Sigma-Aldrich®
Urethane
Formula: C₃H₇NO₂
Boiling Pt: 182 °C (1013 hPa)
Melting Pt: 48…50 °C
Density: 1.11 g/cm³ (20 °C)
Flash Pt: 92 °C
MDL Number: MFCD00007966
CAS Number: 51-79-6
EINECS: 200-123-1
UN: 2757
ADR: 6.1,II
Merck Index: 12,10013