Methyl carbamate ≥99%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: Carbamic acid methyl ester
A16523.18 A16523.22 A16523.36
A16523.18EA 27.1 EUR
A16523.18 A16523.22 A16523.36
Methyl carbamate ≥99%
Methyl carbamate
Purity: 99% CAS No.: 598-55-0 Molecular Formula: H2NCO2CH3 Formula Weight: 75.07 Flash Point: 100°(212°F) Melting Point: 54-56° Boiling Point: 176-178° Beilstein Reference: 635779 Merck Reference: 14,6036 MDL No.: MFCD00007964
Formula: C₂H₅NO₂
Boiling Pt: 176…178 °C
Melting Pt: 54…56 °C
Density: 1.140
Flash Pt: >100 °C (212 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007964
CAS Number: 598-55-0
EINECS: 209-939-2
Merck Index: 14,6036