Silver diethyldithiocarbamate, Sigma-Aldrich®

Supplier: SIGMA ALDRICH MICROSCOPY
Warning

D3132-25G D3132-5G
D3132-25GEA 286 EUR
D3132-25G D3132-5G
Silver diethyldithiocarbamate, Sigma-Aldrich®
Silver diethyldithiocarbamate

Spin trap used in conjunction with Fe²⁺ to detect NO in brain, kidney, liver, and other tissues. Simultaneous direct measurement of NO* and pO₂ has been reported. A copper chelator, it has been used to quantitate copper in biological materials by ESR and is an inhibitor of superoxide dismutase, ascorbate oxidase, and other enzymes. Also used for colorimetric determination of small quantities of arsenic. Silver diethyldithiocarbamate has been used in cell culture. Also used for colorimetric determination of small quantities of arsenic. Spin trap used in conjunction with Fe²⁺ to detect NO in brain, kidney, liver, and other tissues. Simultaneous direct measurement of NO* and pO₂ has been reported. A copper chelator, it has been used to quantitate copper in biological materials by ESR and is an inhibitor of superoxide dismutase, ascorbate oxidase, and other enzymes.


  • Inhibits induction of macrophage nitric oxide synthase
  • Silver diethyldithiocarbamate (AgDDC) is used to estimate the level of arsenic in human
  • Inhibits induction of macrophage nitric oxide synthase

Formula: (C₂H₅)₂NCSSAg
Melting Pt: 172…175 °C (lit.)
Storage Temperature: Refrigerator
MDL Number: MFCD00004929
CAS Number: 1470-61-7
EINECS: 216-003-7
UN: 3077
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c07412a6ac577902b87e980dc94c68b5023241ea&vid=1008727afdc6133fab6eb361e7594c49f93728c2&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR