Silver diethyldithiocarbamate, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: N,N-Diethyldithiocarbamic acid silver salt, Diethyldithiocarbamic acid silver salt, DETC, (Diethyldithiocarbamato-S,S')silver
M-Clarity™ quality level = MQ200
D93503-25G D93503-5G
SIALD93503-25GEA 288 EUR
SIALD93503-25G SIALD93503-5G
Silver diethyldithiocarbamate, Sigma-Aldrich®
Silver diethyldithiocarbamate
Formula: (C₂H₅)₂NCSSAg
MW: 256.14 g/mol
Melting Pt: 176…178.5 °C
MDL Number: MFCD00004929
CAS Number: 1470-61-7
EINECS: 216-003-7
UN: 3077
ADR: 9 III