3-(Dimethylamino)-1-propanol, Sigma-Aldrich®

Supplier: Merck

D144401-100ML
SIALD144401-100MLEA 30.7 EUR
SIALD144401-100ML
3-(Dimethylamino)-1-propanol, Sigma-Aldrich®
3-(Dimethylamino)-1-propanol
Formula: C₅H₁₃NO
Boiling Pt: 163 °C (1013 hPa)
Melting Pt: –35 °C
Density: 0.8878 g/cm³ (20 °C)
Flash Pt: 58 °C
MDL Number: MFCD00002945
CAS Number: 3179-63-3
EINECS: 221-659-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR