Acetal, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 1,1-Diethoxyethane, Acetaldehyde diethyl acetal
M-Clarity™ quality level = MQ200
A902-100ML A902-500ML A902-5ML
SIALA902-100MLEA 59.6 EUR
SIALA902-100ML SIALA902-500ML SIALA902-5ML
Acetal, Sigma-Aldrich®
Acetal
Formula: C₆H₁₄O₂
MW: 118.18 g/mol
Boiling Pt: 102.7
Melting Pt: –100 °C
Density: 0.83 g/cm³ (20 °C)
Flash Pt: –21 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009243
CAS Number: 105-57-7
EINECS: 203-310-6
UN: 1088
ADR: 3 II
Merck Index: 13,00040