Dimethyl acetal, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Acetaldehyde dimethyl acetal, 1,1-Dimethoxyethane
M-Clarity™ quality level = MQ100
385395-100ML 385395-25ML
SIAL385395-100MLEA 200 EUR
SIAL385395-100ML SIAL385395-25ML
Dimethyl acetal, Sigma-Aldrich®
Dimethyl acetal
Formula: C₄H₁₀O₂
MW: 90.12 g/mol
Boiling Pt: 64
Melting Pt: –113 °C
Density: 0.8505 g/cm³ (20 °C)
MDL Number: MFCD00008493
CAS Number: 534-15-6
EINECS: 208-589-8
UN: 2377
ADR: 3 II
Merck Index: 13,03253